AMBH324A598F Ambeed, Inc.

Hydroquinidine 1,4-phthalazinediyl ether

  • MDL number MFCD00198107
  • Cas Number : 140853-10-7
  • Molecular Weight : 778.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/140853-10-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/140853-10-7.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart