AMBH324A5993 Ambeed, Inc.

(S)-2,2-Diphenyl-2-(pyrrolidin-3-yl)acetamide (2R,3R)-2,3-dihydroxysuccinate

  • MDL number MFCD09750973
  • Cas Number : 134002-26-9
  • Molecular Weight : 430.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/134002-26-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/134002-26-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart