AMBH324A599D Ambeed, Inc.

L-Rhamnose Monohydrate

  • MDL number MFCD00149363
  • Cas Number : 10030-85-0
  • Molecular Weight : 182.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/10030-85-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/10030-85-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart