AMBH324A599F Ambeed, Inc.

RAD51 inhibitor B02

  • MDL number MFCD03294274
  • Cas Number : 1290541-46-6
  • Molecular Weight : 339.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1290541-46-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1290541-46-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart