AMBH324A59A0 Ambeed, Inc.

Fradiomycin Sulfate

  • MDL number MFCD00037007
  • Cas Number : 1405-10-3
  • Molecular Weight : 712.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 650
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1405-10-3.html

Safety Information

signalword Warning
pcodes P422-P280-P305+P351+P338-P231+P232
MSDS https://www.ambeed.com/products/1405-10-3.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart