AMBH324A59A2 Ambeed, Inc.

SBI-0640756

  • MDL number MFCD30343858
  • Cas Number : 1821280-29-8
  • Molecular Weight : 404.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1821280-29-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1821280-29-8.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart