AMBH324A59A9 Ambeed, Inc.

2'-Fluoro-2'-deoxyadenosine

  • MDL number MFCD09750859
  • Cas Number : 64183-27-3
  • Molecular Weight : 269.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/64183-27-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/64183-27-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart