AMBH324A59BB Ambeed, Inc.

(R)-3-(5-(2-Aminopropyl)-7-cyanoindolin-1-yl)propyl benzoate (2R,3R)-2,3-dihydroxysuccinate

  • MDL number MFCD16038449
  • Cas Number : 239463-85-5
  • Molecular Weight : 513.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/239463-85-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/239463-85-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart