AMBH324A59CF Ambeed, Inc.

H-Lys(Me)-OH.HCl

  • MDL number MFCD00012621
  • Cas Number : 7622-29-9
  • Molecular Weight : 196.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/7622-29-9.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/7622-29-9.html
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart