AMBH324A59D0 Ambeed, Inc.

(S)-2-Amino-2-(3-fluoro-5-methoxyphenyl)ethanol

  • MDL number MFCD09415636
  • Cas Number : 1212981-84-4
  • Molecular Weight : 185.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1212981-84-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1212981-84-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart