AMBH324A59D3 Ambeed, Inc.

(S)-Ethyl 2-benzamido-3-(4-nitrophenyl)propanoate

  • MDL number MFCD19690954
  • Cas Number : 58816-69-6
  • Molecular Weight : 342.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/58816-69-6.html

Safety Information

signalword Danger
pcodes P273
MSDS https://www.ambeed.com/products/58816-69-6.html
pictograms GHS07,GHS09
hstatements H302-H400
Availability details for
Quantity
  • Availability
  • Add to Cart