AMBH324A59D5 Ambeed, Inc.

Pimecrolimus

  • MDL number MFCD00901792
  • Cas Number : 137071-32-0
  • Molecular Weight : 810.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/137071-32-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/137071-32-0.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart