AMBH324A59D6 Ambeed, Inc.

(E)-(4-Chlorostyryl)boronic acid

  • MDL number MFCD02093767
  • Cas Number : 154230-29-2
  • Molecular Weight : 182.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/154230-29-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/154230-29-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart