AMBH324AB78E Ambeed, Inc.

2-(Boc-amino)furan

  • MDL number MFCD02682023
  • Cas Number : 56267-47-1
  • Molecular Weight : 183.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98°C at N/A mmHg
Purity 97
Flash Point 78°C
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 205.3°C at 760 mmHg
Product Link https://www.ambeed.com/products/56267-47-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/56267-47-1.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart