AMBH324AB79B Ambeed, Inc.

3-(Benzyloxy)-4-methylaniline

  • MDL number MFCD04973042
  • Cas Number : 219492-12-3
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Flash Point 184.1°C
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Physical Form Solid or liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 369.3°C at 760 mmHg
Product Link https://www.ambeed.com/products/219492-12-3.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/219492-12-3.html
pictograms GHS07
hstatements H302-H312-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart