AMBH324AB7A6 Ambeed, Inc.

2-(Trifluoromethyl)benzohydrazide

  • MDL number MFCD00221473
  • Cas Number : 344-95-6
  • Molecular Weight : 204.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-134°C at N/A mmHg
Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/344-95-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/344-95-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart