AMBH324AB7B1 Ambeed, Inc.

4-Methoxybenzofuran-3(2H)-one

  • MDL number MFCD12828273
  • Cas Number : 7169-35-9
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 108.7°C
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 261.7°C at 760 mmHg
Product Link https://www.ambeed.com/products/7169-35-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/7169-35-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart