AMBH324AB7B5 Ambeed, Inc.

5-(Methoxycarbonyl)-2-nitrobenzoic acid

  • MDL number MFCD08690063
  • Cas Number : 76143-33-4
  • Molecular Weight : 225.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 219.4°C
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 439.2°40.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/76143-33-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/76143-33-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart