AMBH324AB7B7 Ambeed, Inc.

4-Chloro-7-(3-chloropropoxy)-6-methoxyquinoline-3-carbonitrile

  • MDL number MFCD09833974
  • Cas Number : 214470-68-5
  • Molecular Weight : 311.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 243°C
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 478.1°C at 760 mmHg
Product Link https://www.ambeed.com/products/214470-68-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/214470-68-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart