AMBH324AB7BB Ambeed, Inc.

5-Bromo-2-fluoro-3-pyridylboronic

  • MDL number MFCD04039317
  • Cas Number : 501435-91-2
  • Molecular Weight : 219.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300°C
Purity 97
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 357.1°C at 760 mmHg
Product Link https://www.ambeed.com/products/501435-91-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/501435-91-2.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart