AMBH324AB7C2 Ambeed, Inc.

Ethyl 3-(2,6-Difluorophenyl)-3-oxopropionate

  • MDL number MFCD11041490
  • Cas Number : 97305-12-9
  • Molecular Weight : 228.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 266.0°25.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/97305-12-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/97305-12-9.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart