AMBH324AB7D9 Ambeed, Inc.

Zinc stearate

  • MDL number MFCD00013031
  • Cas Number : 557-05-1
  • Molecular Weight : 632.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-128°C at N/A mmHg
Purity 12.514
Flash Point 162.4°C
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 359.4°C at 760 mmHg
Product Link https://www.ambeed.com/products/557-05-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/557-05-1.html
pictograms GHS07
hstatements H302-H315-H319-H335-H413
Availability details for
Quantity
  • Availability
  • Add to Cart