AMBH324AB7DA Ambeed, Inc.

Methyl N-Cbz-4-piperidineacetate

  • MDL number MFCD02179006
  • Cas Number : 170737-53-8
  • Molecular Weight : 291.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 197.5°C
Storage temp. Sealed in dry,Room Temperature
Physical Form Colorless or white to brown liquid or solid
Shipping Temperature Normal
Country of Origin China
Boiling point 402.9°28.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/170737-53-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/170737-53-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart