AMBH324AB7DD Ambeed, Inc.

1,3-Bis(chloromethyl)tetramethyldisiloxane

  • MDL number MFCD00000877
  • Cas Number : 2362-10-9
  • Molecular Weight : 231.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -90°C at N/A mmHg
Purity 95
Flash Point 63.3°C
Storage temp. Inert atmosphere,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 204.7°C at 760 mmHg
Product Link https://www.ambeed.com/products/2362-10-9.html

Safety Information

signalword Warning
pcodes P210-P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313-P370+P378-P403+P235-P501
MSDS https://www.ambeed.com/products/2362-10-9.html
pictograms GHS07
hstatements H227-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart