AMBH324AB7E6 Ambeed, Inc.

5-Methoxy-1,2,3,4-tetrahydronaphthalen-2-amine

  • MDL number MFCD09026453
  • Cas Number : 4018-91-1
  • Molecular Weight : 177.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 147.4°C
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Physical Form Solid or liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 309.1°C at 760 mmHg
Product Link https://www.ambeed.com/products/4018-91-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/4018-91-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart