AMBH324AB7ED Ambeed, Inc.

5-(3,4-Dimethoxybenzyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD00980511
  • Cas Number : 313957-85-6
  • Molecular Weight : 251.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 220.2°C
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 440.5°C at 760 mmHg
Product Link https://www.ambeed.com/products/313957-85-6.html

Safety Information

signalword Warning
pcodes P305+P351+P338-P280
MSDS https://www.ambeed.com/products/313957-85-6.html
pictograms GHS07
hstatements H319-H317
Availability details for
Quantity
  • Availability
  • Add to Cart