AMBH324AB7EE Ambeed, Inc.

2-Bromo-4-(trifluoromethyl)thiazole-5-carboxylic acid

  • MDL number MFCD09264552
  • Cas Number : 162651-07-2
  • Molecular Weight : 276.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-132°C at N/A mmHg
Purity 97
Flash Point 150.927°C
Storage temp. Inert atmosphere,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin China
Boiling point 325.946°C at 760 mmHg
Product Link https://www.ambeed.com/products/162651-07-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/162651-07-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart