AMBH324AB7F1 Ambeed, Inc.

2-(Boc-aminomethyl)benzyl Alcohol

  • MDL number MFCD22418063
  • Cas Number : 1333114-86-5
  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1333114-86-5.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312-P330
MSDS https://www.ambeed.com/products/1333114-86-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart