AMBH324AB7F7 Ambeed, Inc.

Benzo[d]thiazole-6-sulfonyl chloride

  • MDL number MFCD03086100
  • Cas Number : 181124-40-3
  • Molecular Weight : 233.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-115°C
Purity 96
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 376.9°C at 760 mmHg
Product Link https://www.ambeed.com/products/181124-40-3.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/181124-40-3.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart