AMBH324AB809 Ambeed, Inc.

(3-(Difluoromethoxy)phenyl)methanamine

  • MDL number MFCD00236230
  • Cas Number : 244022-71-7
  • Molecular Weight : 173.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 226.8°C at 760 mmHg
Product Link https://www.ambeed.com/products/244022-71-7.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/244022-71-7.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart