AMBH324AB814 Ambeed, Inc.

2-(Boc-amino)indane-2-carboxylic Acid

  • MDL number MFCD00800597
  • Cas Number : 71066-00-7
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 229.9°C
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 456.6°C at 760 mmHg
Product Link https://www.ambeed.com/products/71066-00-7.html

Safety Information

signalword Danger
pcodes P233-P260-P261-P264-P270-P271-P273-P280-P301+P312-P302+P352-P304-P304+P340-P305+P351+P338-P312-P321-P330-P332+P313-P337+P313-P340-P362-P391-P403-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/71066-00-7.html
pictograms GHS07,GHS09
hstatements H302-H315-H319-H332-H335-H410
Availability details for
Quantity
  • Availability
  • Add to Cart