AMBH324AB81E Ambeed, Inc.

4-(Piperidin-1-ylmethyl)phenol

  • MDL number MFCD18453499
  • Cas Number : 73152-41-7
  • Molecular Weight : 191.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 306.7°17.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/73152-41-7.html

Safety Information

signalword Warning
pcodes P305+P351+P338-P280
MSDS https://www.ambeed.com/products/73152-41-7.html
pictograms GHS07
hstatements H319-H317
Availability details for
Quantity
  • Availability
  • Add to Cart