AMBH324AB828 Ambeed, Inc.

2-Bromo-5-methoxypyrimidine

  • MDL number MFCD12025462
  • Cas Number : 1209459-99-3
  • Molecular Weight : 189.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 98
Flash Point 131.4°C
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 293.7°C at 760 mmHg
Product Link https://www.ambeed.com/products/1209459-99-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1209459-99-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart