AMBH324AB83B Ambeed, Inc.

1-Butyl-3-methylpyridinium Chloride

  • MDL number MFCD08458942
  • Cas Number : 125652-55-3
  • Molecular Weight : 185.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/125652-55-3.html

Safety Information

signalword Warning
pcodes P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313
MSDS https://www.ambeed.com/products/125652-55-3.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart