AMBH324AB858 Ambeed, Inc.

2-(1-Benzylpiperidin-4-yl)-2-propanol

  • MDL number MFCD19440844
  • Cas Number : 299428-04-9
  • Molecular Weight : 233.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 139.059°C
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 331.052°C at 760 mmHg
Product Link https://www.ambeed.com/products/299428-04-9.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/299428-04-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart