AMBH324AB85C Ambeed, Inc.

2-Chloro-N-[5-(dimethylamino)-2-nitrophenyl]-5-nitrobenzamide

  • MDL number MFCD27918643
  • Cas Number : 1079264-82-6
  • Molecular Weight : 364.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215-217°C at N/A mmHg
Purity 95
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 494.2°45.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/1079264-82-6.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/1079264-82-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart