AMBH324AB87F Ambeed, Inc.

N-[3-(Trifluoromethyl)benzyl]ethylamine

  • MDL number MFCD09935363
  • Cas Number : 14355-04-5
  • Molecular Weight : 203.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 74.6°C
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 199.7°C at 760 mmHg
Product Link https://www.ambeed.com/products/14355-04-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/14355-04-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart