AMBH324AB882 Ambeed, Inc.

3-Hydroxy-2-nitrobenzonitrile

  • MDL number MFCD09834347
  • Cas Number : 129298-23-3
  • Molecular Weight : 164.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 310.1°32.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/129298-23-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/129298-23-3.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart