AMBH324AB884 Ambeed, Inc.

2-Chloro-7,8-dihydroquinolin-5(6H)-one

  • MDL number MFCD08692066
  • Cas Number : 124467-36-3
  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119-123°C
Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 275.3°C at 760 mmHg
Product Link https://www.ambeed.com/products/124467-36-3.html

Safety Information

signalword Danger
pcodes P261-P273-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/124467-36-3.html
pictograms GHS05,GHS07,GHS09
hstatements H302-H315-H318-H335-H410
Availability details for
Quantity
  • Availability
  • Add to Cart