AMBH324AB886 Ambeed, Inc.

1-(Bromomethyl)-2-fluoro-3-methoxybenzene

  • MDL number MFCD09743597
  • Cas Number : 447463-56-1
  • Molecular Weight : 219.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 98
Flash Point 123.2°C
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 247.5°C at 760 mmHg
Product Link https://www.ambeed.com/products/447463-56-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/447463-56-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart