AMBH324AB896 Ambeed, Inc.

1-Benzyl-1H-1,2,3-triazole

  • MDL number MFCD06251908
  • Cas Number : 4368-68-7
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 97
Flash Point 141.4°C
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 310.3°C at 760 mmHg
Product Link https://www.ambeed.com/products/4368-68-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/4368-68-7.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart