AMBH324AB898 Ambeed, Inc.

Potassium trifluoro(3-nitrophenyl)borate

  • MDL number MFCD03701627
  • Cas Number : 192863-40-4
  • Molecular Weight : 229.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 260°C at N/A mmHg
Purity 95
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/192863-40-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/192863-40-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart