AMBH324AB89A Ambeed, Inc.

3-Chloro-2-nitrobenzaldehyde

  • MDL number MFCD06797953
  • Cas Number : 22233-52-9
  • Molecular Weight : 185.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 313.6°27.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/22233-52-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/22233-52-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart