AMBH324AB8AB Ambeed, Inc.

1-Naphthamide

  • MDL number MFCD00014314
  • Cas Number : 2243-81-4
  • Molecular Weight : 171.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201-204°C at N/A mmHg
Purity 98
Flash Point 196.6°C
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 401.5°C at 760 mmHg
Product Link https://www.ambeed.com/products/2243-81-4.html

Safety Information

signalword Warning
pcodes P261-P301+P312-P302+P352-P304+P340-P305+P351+P338
MSDS https://www.ambeed.com/products/2243-81-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart