AMBH324AB8AF Ambeed, Inc.

3-Chloro-4-(trifluoromethyl)benzoic acid

  • MDL number MFCD04972755
  • Cas Number : 115754-20-6
  • Molecular Weight : 224.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-161°C at N/A mmHg
Purity 98
Flash Point 124.2°C
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 281.7°C at 760 mmHg
Product Link https://www.ambeed.com/products/115754-20-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/115754-20-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart