AMBH324AB8B3 Ambeed, Inc.

3-Amino-5-methylpyridazine

  • MDL number MFCD10698051
  • Cas Number : 144294-43-9
  • Molecular Weight : 109.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 320.8°C at 760 mmHg
Product Link https://www.ambeed.com/products/144294-43-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/144294-43-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart