AMBH324AB8C0 Ambeed, Inc.

2'-Hydroxy-3'-methoxyacetophenone

  • MDL number MFCD12913657
  • Cas Number : 703-98-0
  • Molecular Weight : 166.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54°C
Purity 97
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 247.2°C at 760 mmHg
Product Link https://www.ambeed.com/products/703-98-0.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/703-98-0.html
pictograms GHS07
hstatements H302-H319
Availability details for
Quantity
  • Availability
  • Add to Cart