AMBH324AB8CD Ambeed, Inc.

2-(Bromomethyl)-1-methoxy-4-nitrobenzene

  • MDL number MFCD00007329
  • Cas Number : 3913-23-3
  • Molecular Weight : 246.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76-78°C
Synonym N/A
Purity 95
Flash Point 164°C
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 348.7°C at 760 mmHg
Product Link https://www.ambeed.com/products/3913-23-3.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/3913-23-3.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart