AMBH324AB8CF Ambeed, Inc.

Thiazolo[4,5-b]pyridin-2-amine

  • MDL number MFCD11847509
  • Cas Number : 13575-41-2
  • Molecular Weight : 151.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118.1°C
Purity 97
Flash Point 153°C
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 329°C at 760 mmHg
Product Link https://www.ambeed.com/products/13575-41-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/13575-41-2.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart