AMBH324AB8D7 Ambeed, Inc.

Mpr(Bzl)-OH

  • MDL number MFCD00020521
  • Cas Number : 2899-66-3
  • Molecular Weight : 196.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Benzylthio)propanoic acid
Purity 98
Flash Point 166.4°C
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 351.6°C at 760 mmHg
Product Link https://www.ambeed.com/products/2899-66-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/2899-66-3.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart